Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109045.1 | NP_001102515.1 | 2 - 3 | 410 | 135 | |
NM_007474.2 | NP_031500.1 | 2 - 3 | 413 | 135 | ||
GenBank mRNA | AF018952.1 | - | 2 - 3 | 395 | 135 | |
BC010982.1 | - | 2 - 3 | 355 | 4224850 | 135 | |
CF617211.1 | - | 2 - 3 | 316 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |