Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144887.4 | NP_659136.1 | 6 - 7 | 1900 | 106 | |
GenBank mRNA | AK028737.1 | - | 3 - 4 | 466 | 106 | |
AK082513.1 | - | 6 - 7 | 1825 | 106 | ||
AK145086.1 | - | 6 - 7 | 1827 | 106 | ||
AK173251.1 | - | 6 - 7 | 1855 | 106 | ||
AY894891.1 | - | 6 - 7 | 1892 | 106 | ||
BC020051.1 | - | 5 - 6 | 870 | 3987696 | 106 | |
BC027047.1 | - | 2 - 3 | 134 | 106 | ||
BC065155.1 | - | 6 - 7 | 1770 | 5706087 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |