Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027363.1 | NP_081639.1 | 1 - 2 | 284 | 69 | |
GenBank mRNA | AK008392.1 | - | 1 - 2 | 118 | 69 | |
BC119202.1 | - | 1 - 2 | 284 | 8733951 | 69 | |
BC119204.1 | - | 1 - 2 | 284 | 8733953 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |