Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027363.1 | NP_081639.1 | 3 - 4 | 439 | 64 | |
GenBank mRNA | AK008392.1 | - | 3 - 4 | 273 | 64 | |
BC119202.1 | - | 3 - 4 | 439 | 8733951 | 64 | |
BC119204.1 | - | 3 - 4 | 439 | 8733953 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |