Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ern2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012016.2 | NP_036146.2 | 4 - 5 | 367 | 76 | |
GenBank mRNA | AK139291.1 | - | 4 - 5 | 359 | 76 | |
AK153659.1 | - | 4 - 5 | 357 | 76 | ||
BC108999.2 | - | 4 - 5 | 326 | 40048029 | 76 | |
BC109000.1 | - | 4 - 5 | 326 | 40048030 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |