Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172409.2 | NP_765997.2 | 13 - 14 | 1590 | 89 | |
GenBank mRNA | AF513716.1 | - | 1 - 2 | 92 | 89 | |
BM114825.1 | - | 1 - 2 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |