Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101488.1 | NP_001094958.1 | 5 - 6 | 1035 | 86 | |
GenBank mRNA | AK082731.1 | - | 5 - 6 | 1038 | 86 | |
BM950164.1 | - | 3 - 4 | 425 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |