Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145824.1 | NP_001139296.1 | 3 - 4 | 1770 | 63 | |
NM_010434.2 | NP_034564.2 | 3 - 4 | 1770 | 63 | ||
GenBank mRNA | AF170305.1 | - | 2 - 3 | 1234 | 63 | |
AF305238.1 | - | 2 - 3 | 1232 | 63 | ||
AK140130.1 | - | 3 - 4 | 1873 | 63 | ||
BC131663.1 | - | 3 - 4 | 1401 | 63 | ||
BC131664.1 | - | 3 - 4 | 1401 | 40110751 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |