Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081117.2 | NP_001074586.2 | 3 - 4 | 525 | 82 | |
GenBank mRNA | AK052772.1 | - | 3 - 4 | 445 | 82 | |
AK143677.1 | - | 3 - 4 | 564 | 82 | ||
X82786.1 | - | 3 - 4 | 630 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |