Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mki67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081117.2 | NP_001074586.2 | 6 - 7 | 1605 | 95 | |
GenBank mRNA | AK052772.1 | - | 6 - 7 | 1525 | 95 | |
AK082099.1 | - | 6 - 7 | 1524 | 95 | ||
AK143677.1 | - | 6 - 7 | 1644 | 95 | ||
X82786.1 | - | 6 - 7 | 1713 | 95 |
Set Membership: |
![]() ![]() ![]() ![]() |