Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk32c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162540.1 | NP_001156012.1 | 8 - 9 | 828 | 146 | |
NM_021302.3 | NP_067277.2 | 8 - 9 | 1128 | 146 | ||
GenBank mRNA | AB041542.1 | - | 8 - 9 | 1108 | 146 | |
AK046439.1 | - | 8 - 9 | 1106 | 146 | ||
BC026457.1 | - | 8 - 9 | 1113 | 4481218 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |