Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011538.2 | NP_035668.2 | 1 - 2 | 138 | 72 | |
GenBank mRNA | AY654733.1 | - | 1 - 2 | 114 | 72 | |
BC140951.1 | - | 1 - 2 | 138 | 9055563 | 72 | |
BF607954.1 | - | 1 - 2 | 173 | 72 | ||
U57331.1 | - | 1 - 2 | 138 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |