Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139559.1 | - | 1 - 2 | 73 | 96 | |
AK145816.1 | - | 1 - 2 | 78 | 96 | ||
AK151203.1 | - | 1 - 2 | 130 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |