Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030697.2 | NP_109622.1 | 5 - 6 | 1647 | 80 | |
GenBank mRNA | AK002623.1 | - | 5 - 6 | 1648 | 80 | |
AK132011.1 | - | 5 - 6 | 1669 | 80 | ||
KJ650203.1 | - | 4 - 5 | 1578 | 80 | ||
KJ663713.1 | - | 4 - 5 | 1419 | 80 | ||
KJ663714.1 | - | 5 - 6 | 1440 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |