Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030697.2 | NP_109622.1 | 8 - 9 | 2097 | 90 | |
GenBank mRNA | AK002623.1 | - | 8 - 9 | 2098 | 90 | |
AK132011.1 | - | 8 - 9 | 2119 | 90 | ||
BC037741.1 | - | 3 - 4 | 231 | 90 | ||
BC061055.1 | - | 3 - 4 | 243 | 90 | ||
BC115933.1 | - | 2 - 3 | 201 | 90 | ||
KJ650203.1 | - | 7 - 8 | 2028 | 90 | ||
KJ663713.1 | - | 7 - 8 | 1869 | 90 | ||
KJ663714.1 | - | 8 - 9 | 1890 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |