Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172620.3 | NP_766208.2 | 1 - 2 | 304 | 113 | |
GenBank mRNA | AK042520.1 | - | 1 - 2 | 258 | 113 | |
AK051732.1 | - | 1 - 2 | 375 | 113 | ||
AK052523.1 | - | 1 - 2 | 305 | 113 | ||
AX775735.1 | - | 1 - 2 | 116 | 113 | ||
BC063329.1 | - | 1 - 2 | 271 | 30251096 | 113 | |
KJ650214.1 | - | 1 - 2 | 93 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |