Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010724.2 | NP_034854.2 | 3 - 4 | 607 | 63 | |
GenBank mRNA | AK031801.1 | - | 3 - 4 | 432 | 63 | |
AK050627.1 | - | 3 - 4 | 435 | 63 | ||
AK146182.1 | - | 3 - 4 | 454 | 63 | ||
AK150674.1 | - | 3 - 4 | 435 | 63 | ||
AK159971.1 | - | 3 - 4 | 424 | 63 | ||
AK169753.1 | - | 2 - 3 | 880 | 63 | ||
BC013785.1 | - | 3 - 4 | 431 | 2654130 | 63 | |
KJ650228.1 | - | 3 - 4 | 403 | 63 | ||
U22031.1 | - | 3 - 4 | 403 | 63 | ||
U22032.1 | - | 3 - 4 | 403 | 63 | ||
U22033.1 | - | 3 - 4 | 403 | 63 | ||
U22034.1 | - | 3 - 4 | 403 | 63 | ||
U22035.1 | - | 3 - 4 | 403 | 63 | ||
X64449.1 | - | 3 - 4 | 422 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |