Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079932.3 | NP_001073401.1 | 4 - 5 | 843 | 75 | |
GenBank mRNA | AB231474.1 | - | 4 - 5 | 747 | 75 | |
BC067209.1 | - | 4 - 5 | 409 | 75 | ||
BC147789.1 | - | 4 - 5 | 843 | 9087507 | 75 | |
BC158111.1 | - | 4 - 5 | 828 | 9088185 | 75 | |
BC158120.1 | - | 4 - 5 | 828 | 9088192 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |