Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053194.4 | NP_444424.1 | 8 - 9 | 1424 | 66 | |
GenBank mRNA | AF288813.1 | - | 8 - 9 | 1423 | 66 | |
AK031466.1 | - | 8 - 9 | 1371 | 66 | ||
AK080192.1 | - | 8 - 9 | 1367 | 66 | ||
AK155282.1 | - | 8 - 9 | 1399 | 66 | ||
AK169367.1 | - | 8 - 9 | 1388 | 66 | ||
BC022917.1 | - | 8 - 9 | 1401 | 4460521 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |