Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013819.2 | NP_038847.1 | 4 - 5 | 924 | 103 | |
GenBank mRNA | AB267096.2 | - | 3 - 4 | 713 | 103 | |
BC031984.1 | - | 4 - 5 | 915 | 4007538 | 103 | |
U18797.1 | - | 4 - 5 | 896 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |