Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026646.3 | NP_080922.1 | 1 - 2 | 304 | 121 | |
GenBank mRNA | BC050887.1 | - | 1 - 2 | 257 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |