Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029935.5 | NP_084211.2 | 2 - 3 | 1103 | 76 | |
GenBank mRNA | AB187269.1 | - | 1 - 2 | 543 | 76 | |
AF320786.1 | - | 1 - 2 | 594 | 76 | ||
AK019474.1 | - | 2 - 3 | 1058 | 76 | ||
AK035637.1 | - | 2 - 3 | 1191 | 76 | ||
AK082318.1 | - | 2 - 3 | 1169 | 76 | ||
AK083614.1 | - | 2 - 3 | 1103 | 76 | ||
AK122324.1 | - | 2 - 3 | 1404 | 76 | ||
BC031443.1 | - | 2 - 3 | 1060 | 4240286 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |