Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029935.5 | NP_084211.2 | 3 - 4 | 1444 | 95 | |
GenBank mRNA | AB187269.1 | - | 2 - 3 | 884 | 95 | |
AF320786.1 | - | 2 - 3 | 935 | 95 | ||
AK019474.1 | - | 3 - 4 | 1399 | 95 | ||
AK031745.1 | - | 1 - 2 | 145 | 95 | ||
AK035637.1 | - | 3 - 4 | 1532 | 95 | ||
AK043803.1 | - | 1 - 2 | 292 | 95 | ||
AK082318.1 | - | 3 - 4 | 1510 | 95 | ||
AK083614.1 | - | 3 - 4 | 1444 | 95 | ||
AK122324.1 | - | 3 - 4 | 1745 | 95 | ||
BC031443.1 | - | 3 - 4 | 1401 | 4240286 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |