Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuzd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008411.3 | NP_032437.3 | 6 - 7 | 1241 | 68 | |
GenBank mRNA | BC140992.1 | - | 6 - 7 | 1176 | 9055604 | 68 |
U69699.2 | - | 6 - 7 | 1240 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |