Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009516.3 | NP_033542.2 | 3 - 4 | 1133 | 86 | |
GenBank mRNA | AK011212.1 | - | 3 - 4 | 909 | 86 | |
AK039572.1 | - | 3 - 4 | 449 | 86 | ||
BC006852.1 | - | 3 - 4 | 1152 | 3156953 | 86 | |
D30743.1 | - | 3 - 4 | 1134 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |