Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053247.4 | NP_444477.2 | 4 - 5 | 782 | 85 | |
GenBank mRNA | AJ311501.1 | - | 4 - 5 | 937 | 85 | |
AK004726.2 | - | 4 - 5 | 782 | 85 | ||
AK160857.1 | - | 3 - 4 | 520 | 85 | ||
BC038653.1 | - | 4 - 5 | 888 | 2936961 | 85 | |
BC038892.1 | - | 4 - 5 | 888 | 3372151 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |