Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285507.1 | NP_001272436.1 | 3 - 4 | 344 | 108 | |
NM_001285512.1 | NP_001272441.1 | 3 - 4 | 344 | 108 | ||
NM_009939.3 | NP_034069.2 | 3 - 4 | 344 | 108 | ||
GenBank mRNA | AF071312.1 | - | 3 - 4 | 254 | 108 | |
AF087688.1 | - | 3 - 4 | 251 | 108 | ||
AK010383.1 | - | 3 - 4 | 270 | 108 | ||
AK037757.1 | - | 3 - 4 | 270 | 108 | ||
AK084421.1 | - | 3 - 4 | 265 | 108 | ||
AK132207.1 | - | 3 - 4 | 261 | 108 | ||
AK132776.1 | - | 3 - 4 | 281 | 108 | ||
AK146153.1 | - | 3 - 4 | 255 | 108 | ||
BC023096.1 | - | 3 - 4 | 249 | 3675508 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |