Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144857.1 | NP_659106.1 | 3 - 4 | 268 | 62 | |
GenBank mRNA | AK148164.1 | - | 414 | 62 | ||
AK148237.1 | - | 3 - 4 | 365 | 62 | ||
BC011248.1 | - | 3 - 4 | 268 | 4192711 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |