Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113179.1 | NP_001106650.1 | 11 - 12 | 1360 | 158 | |
NM_009772.2 | NP_033902.2 | 11 - 12 | 1357 | 158 | ||
GenBank mRNA | AF002823.1 | - | 11 - 12 | 1303 | 158 | |
AK042932.1 | - | 11 - 12 | 1358 | 158 | ||
AK144213.1 | - | 11 - 12 | 1369 | 158 | ||
BC029116.1 | - | 11 - 12 | 1326 | 3671932 | 158 | |
U89795.1 | - | 11 - 12 | 1375 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |