Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029963.2 | NP_084239.1 | 3 - 4 | 286 | 62 | |
GenBank mRNA | AB049941.1 | - | 3 - 4 | 286 | 62 | |
AK018829.1 | - | 3 - 4 | 294 | 62 | ||
AK078198.1 | - | 3 - 4 | 294 | 62 | ||
AK083366.1 | - | 3 - 4 | 288 | 62 | ||
AK155101.1 | - | 3 - 4 | 302 | 62 | ||
BC021947.1 | - | 3 - 4 | 284 | 5098933 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |