Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170912.1 | NP_001164383.1 | 13 - 14 | 3113 | 65 | |
NM_001170913.1 | NP_001164384.1 | 13 - 14 | 3151 | 65 | ||
NM_181853.4 | NP_862901.3 | 13 - 14 | 3050 | 65 | ||
GenBank mRNA | AK147559.1 | - | 13 - 14 | 3152 | 65 | |
BC138032.1 | - | 12 - 13 | 2723 | 8861051 | 65 | |
BC145106.1 | - | 12 - 13 | 2723 | 9053654 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |