Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uhmk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010633.3 | NP_034763.3 | 4 - 5 | 1007 | 75 | |
GenBank mRNA | AK013347.1 | - | 4 - 5 | 1012 | 75 | |
AK030152.1 | - | 824 | 75 | |||
BC058732.1 | - | 4 - 5 | 1007 | 6414877 | 75 | |
X82320.1 | - | 3 - 4 | 493 | 75 | ||
Y10725.2 | - | 4 - 5 | 864 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |