Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010476.3 | NP_034606.3 | 3 - 4 | 405 | 76 | |
GenBank mRNA | AK028380.1 | - | 3 - 4 | 361 | 76 | |
AK050211.1 | - | 3 - 4 | 362 | 76 | ||
AK077914.1 | - | 3 - 4 | 403 | 76 | ||
BC011464.1 | - | 3 - 4 | 369 | 76 | ||
Y15733.1 | - | 3 - 4 | 395 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |