Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025867.2 | NP_080143.1 | 2 - 3 | 232 | 119 | |
GenBank mRNA | AK009003.1 | - | 2 - 3 | 234 | 119 | |
AK009855.1 | - | 2 - 3 | 233 | 119 | ||
AY367772.1 | - | 1 - 2 | 168 | 119 | ||
BC010313.1 | - | 1 - 2 | 199 | 2646766 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |