Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300000282.mm.1 |
NM_001163328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000282.mm.1 |
NM_001163328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000282.mm.1 |
NM_178631.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000282.mm.1 |
NM_178631.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000282.mm.1 |
TC0300000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000292.mm.1 |
NM_001163328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000292.mm.1 |
NM_001163328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000292.mm.1 |
NM_178631.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000292.mm.1 |
NM_178631.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000292.mm.1 |
TC0300000078.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000281.mm.1 |
NM_001163328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000281.mm.1 |
NM_001163328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000281.mm.1 |
NM_178631.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000281.mm.1 |
NM_178631.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000281.mm.1 |
TC0300000078.mm.1 |
MTA TC |