Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019634.2 | NP_062608.2 | 2 - 3 | 314 | 58 | |
GenBank mRNA | AF052492.1 | - | 2 - 3 | 262 | 58 | |
AK004776.1 | - | 2 - 3 | 274 | 58 | ||
AK147306.1 | - | 2 - 3 | 276 | 58 | ||
AK147404.1 | - | 2 - 3 | 278 | 58 | ||
AK214746.1 | - | 1 - 2 | 79 | 58 | ||
BC026516.1 | - | 1 - 2 | 113 | 4911317 | 58 | |
BC050153.1 | - | 2 - 3 | 262 | 58 | ||
BC058603.1 | - | 2 - 3 | 270 | 6529011 | 58 | |
BY238972.1 | - | 2 - 3 | 314 | 58 | ||
D26483.1 | - | 2 - 3 | 262 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |