Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008300.3 | NP_032326.3 | 8 - 9 | 1297 | 78 | |
GenBank mRNA | AK053601.1 | - | 8 - 9 | 1231 | 78 | |
AK133772.1 | - | 8 - 9 | 1239 | 78 | ||
AK155305.1 | - | 8 - 9 | 1214 | 78 | ||
AK166102.1 | - | 8 - 9 | 1282 | 78 | ||
AK220167.1 | - | 8 - 9 | 1248 | 78 | ||
BC003770.1 | - | 8 - 9 | 1206 | 3494648 | 78 | |
BC003950.1 | - | 8 - 9 | 1192 | 78 | ||
CN458707.1 | - | 2 - 3 | 239 | 78 | ||
D85904.1 | - | 8 - 9 | 999 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |