Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164190.1 | NP_001157662.1 | 12 - 13 | 1543 | 134 | |
NM_001164191.1 | NP_001157663.1 | 12 - 13 | 1434 | 134 | ||
NM_019926.3 | NP_064310.2 | 12 - 13 | 1423 | 134 | ||
GenBank mRNA | AF073996.1 | - | 12 - 13 | 1424 | 134 | |
AK016445.1 | - | 12 - 13 | 1544 | 134 | ||
AK166452.1 | - | 12 - 13 | 1402 | 134 | ||
AK220524.1 | - | 3544 | 134 | |||
BC051022.1 | - | 12 - 13 | 1392 | 134 | ||
BC066847.1 | - | 12 - 13 | 1379 | 132 | ||
BC090984.1 | - | 12 - 13 | 1401 | 30246206 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |