Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008255.2 | NP_032281.2 | 4 - 5 | 521 | 88 | |
GenBank mRNA | AK159899.1 | - | 3 - 4 | 410 | 88 | |
BC019782.1 | - | 3 - 4 | 397 | 88 | ||
BC085083.1 | - | 4 - 5 | 433 | 5320486 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |