Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271388.1 | NP_001258317.1 | 2 - 3 | 784 | 70 | |
NM_001271389.1 | NP_001258318.1 | 2 - 3 | 784 | 70 | ||
NM_023472.2 | NP_075961.1 | 2 - 3 | 784 | 70 | ||
GenBank mRNA | AB221648.1 | - | 1 - 2 | 287 | 70 | |
AF314031.1 | - | 2 - 3 | 618 | 70 | ||
AK054037.1 | - | 2 - 3 | 589 | 70 | ||
AK139343.1 | - | 2 - 3 | 666 | 70 | ||
AK161446.1 | - | 2 - 3 | 788 | 70 | ||
BC066113.1 | - | 1 - 2 | 321 | 30361246 | 70 | |
BC079615.1 | - | 2 - 3 | 601 | 30361702 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |