Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022885.2 | NP_075023.2 | 4 - 5 | 606 | 114 | |
GenBank mRNA | AF461761.1 | - | 4 - 5 | 505 | 114 | |
AK051183.1 | - | 4 - 5 | 606 | 114 | ||
AK088004.1 | - | 4 - 5 | 521 | 114 | ||
AK090176.1 | - | 4 - 5 | 515 | 114 | ||
AK166006.1 | - | 4 - 5 | 416 | 114 | ||
BC033452.1 | - | 4 - 5 | 505 | 3156622 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |