Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268696 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268691 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268695 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268694 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967439 |
BC016231.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967439 |
BC053382.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967439 |
DV662169.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967439 |
AK078297.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1967439 |
AK154739.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
904557 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
401928 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
438855 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683491 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
778411 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
106823 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295042 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
962776 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
925228 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
672508 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
248791 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
567437 |
AK085908.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
19404 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
473344 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
AK003724.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
AK020868.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
AK044518.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
AK160923.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
BC030179.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886949 |
NM_026546.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
871025 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
711538 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
914817 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
875758 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750239 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828770 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
91305 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
159096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10125 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
136432 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P115922 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166031 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967439 |
BC016231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967439 |
BC053382.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967439 |
DV662169.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967439 |
AK078297.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1967439 |
AK154739.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268695 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268696 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268705 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268712 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268698 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268701 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268700 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268693 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268689 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268686 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268687 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268710 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268706 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268703 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268719 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268682 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268720 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268691 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268684 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29268707 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P115922 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02758942 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07972297 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166031 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
BC016231.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
BC053382.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
DV662169.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
AK154739.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P323435 |
NM_022011.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P291509 |
BC016231.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118775 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115108 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166031 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P115922 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10511623 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10511620 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10511624 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10733844 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10511622 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10733853 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10733850 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268691 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268694 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268695 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29268696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268526 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268535 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268543 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268511 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268517 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268690 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268554 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268705 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268533 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29268536 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268671 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268673 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268603 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268672 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268696 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268690 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10845143 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268654 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268609 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268661 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268686 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268718 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268648 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10755663 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268599 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10486462 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268705 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268601 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11015782 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268660 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268600 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10702583 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268607 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10702582 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268554 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268649 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268650 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268684 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268558 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268698 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10637488 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268647 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268710 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268676 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268668 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268700 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268561 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268665 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268716 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10838447 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268689 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268656 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268682 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268604 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268667 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10931742 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268556 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268703 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268708 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268611 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268680 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268693 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268560 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268658 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268553 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268669 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10511626 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268652 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268701 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268608 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29268707 |
- |
- |