Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011420.2 | NP_035550.1 | 1 - 2 | 113 | 132 | |
GenBank mRNA | AK012698.1 | - | 1 - 2 | 83 | 132 | |
AK144778.1 | - | 1 - 2 | 114 | 132 | ||
AK152495.1 | - | 1 - 2 | 88 | 132 | ||
AK167832.1 | - | 1 - 2 | 111 | 132 | ||
AY821787.1 | - | 1 - 2 | 74 | 132 | ||
BC045158.1 | - | 1 - 2 | 77 | 6486685 | 132 | |
CJ058980.1 | - | 1 - 2 | 114 | 132 | ||
U63294.1 | - | 1 - 2 | 91 | 132 | ||
U77714.1 | - | 1 - 2 | 74 | 132 | ||
Y12835.1 | - | 1 - 2 | 99 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |