Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_001161355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_001161355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_001161356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_001161356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_134249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
NM_134249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024042.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024056.mm.1 |
NM_134249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024056.mm.1 |
NM_134249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024056.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_001161355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_001161355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_001161356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_001161356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_134249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
NM_134249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024040.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024049.mm.1 |
NM_001161355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024049.mm.1 |
NM_001161355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024049.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024052.mm.1 |
NM_134249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024052.mm.1 |
NM_134249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024052.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024050.mm.1 |
NM_001161355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024050.mm.1 |
NM_001161355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024050.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024044.mm.1 |
NM_001161356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024044.mm.1 |
NM_001161356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024044.mm.1 |
TC1100002611.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_001161355.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_001161355.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_001161356.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_001161356.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_134249.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
NM_134249.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100024041.mm.1 |
TC1100002611.mm.1 |
MTA TC |