Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300015534.mm.1 |
NM_001162884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015534.mm.1 |
NM_001162884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015534.mm.1 |
TC0300002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015532.mm.1 |
NM_001162884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015532.mm.1 |
NM_001162884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015532.mm.1 |
TC0300002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015533.mm.1 |
NM_001162884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015533.mm.1 |
NM_001162884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015533.mm.1 |
TC0300002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015535.mm.1 |
NM_001162884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015535.mm.1 |
NM_001162884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015535.mm.1 |
TC0300002056.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300015536.mm.1 |
NM_001162884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015536.mm.1 |
NM_001162884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300015536.mm.1 |
TC0300002056.mm.1 |
MTA TC |