Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200004483.mm.1 |
NM_010431.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004483.mm.1 |
NM_010431.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004483.mm.1 |
TC1200000652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004485.mm.1 |
NM_010431.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004485.mm.1 |
NM_010431.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004485.mm.1 |
TC1200000652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004487.mm.1 |
NM_010431.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004487.mm.1 |
NM_010431.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004487.mm.1 |
TC1200000652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004484.mm.1 |
NM_010431.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004484.mm.1 |
NM_010431.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004484.mm.1 |
TC1200000652.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200004488.mm.1 |
NM_010431.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004488.mm.1 |
NM_010431.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200004488.mm.1 |
TC1200000652.mm.1 |
MTA TC |