Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172578.2 | NP_766166.2 | 5 - 6 | 1429 | 156 | |
GenBank mRNA | AK032698.1 | - | 3 - 4 | 460 | 156 | |
AK035117.1 | - | 5 - 6 | 1428 | 156 | ||
AK220223.1 | - | 4 - 5 | 695 | 156 | ||
BC052175.1 | - | 5 - 6 | 1429 | 6394068 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |