Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293313.1 | NP_001280242.1 | 3 - 4 | 352 | 76 | |
NM_138314.4 | NP_612187.2 | 3 - 4 | 355 | 76 | ||
NM_178071.6 | NP_835172.1 | 3 - 4 | 352 | 76 | ||
GenBank mRNA | AF202048.1 | - | 2 - 3 | 256 | 76 | |
AK043515.1 | - | 3 - 4 | 339 | 76 | ||
AK044962.1 | - | 3 - 4 | 295 | 76 | ||
AK085222.1 | - | 3 - 4 | 300 | 76 | ||
AK133221.1 | - | 823 | 76 | |||
AK144915.1 | - | 3 - 4 | 356 | 76 | ||
AK164652.1 | - | 3 - 4 | 300 | 76 | ||
BC049225.1 | - | 2 - 3 | 283 | 3483725 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |