Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177839.3 | NP_808507.2 | 11 - 12 | 2767 | 85 | |
GenBank mRNA | AF455756.1 | - | 11 - 12 | 2734 | 85 | |
AK048033.1 | - | 4 - 5 | 882 | 85 | ||
BC138335.1 | - | 11 - 12 | 2728 | 8861355 | 85 | |
BC138336.1 | - | 11 - 12 | 2728 | 8861356 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |