Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163516.2 | NP_001156988.1 | 3 - 4 | 649 | 74 | |
NM_021483.4 | NP_067458.2 | 5 - 6 | 759 | 74 | ||
GenBank mRNA | AB032591.1 | - | 5 - 6 | 754 | 74 | |
AK083141.1 | - | 3 - 4 | 541 | 74 | ||
HQ646186.1 | - | 6 - 7 | 978 | 74 | ||
HQ646187.1 | - | 5 - 6 | 912 | 74 | ||
HQ646188.1 | - | 4 - 5 | 807 | 74 | ||
HQ646189.1 | - | 4 - 5 | 840 | 74 | ||
HQ646190.1 | - | 3 - 4 | 735 | 74 | ||
HQ646191.1 | - | 5 - 6 | 1568 | 74 | ||
HQ646192.1 | - | 6 - 7 | 982 | 74 | ||
HQ646193.1 | - | 6 - 7 | 888 | 74 | ||
HQ646194.1 | - | 5 - 6 | 783 | 74 | ||
HQ646195.1 | - | 6 - 7 | 756 | 74 | ||
HQ646196.1 | - | 5 - 6 | 651 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |